Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB2514

Sigma-Aldrich

Anti-Integrin α5β1 Antibody, clone BMB5

ascites fluid, clone BMB5, Chemicon®

Synonym(s):

VLA-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

BMB5, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG3

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGA5(3678)

Specificity

Antibody MAB2514 was raised against mouse integrin VLA-5 (alpha5beta1), a fibronectin receptor. The antibody was prepared by fusion of spleen cells from a Fisher rat immunized with mouse embryonal carcinoma P19 cells with myeloma P3X63Ag9.653 cells.Specificity of the antibody was verified by its ability to immunoprecipitate alpha5beta1 heterodimers from (125)-I-surface labeled cells, reciprocal depletion of VLA-5 antigen from cell lysate with antiserum against the cytoplasmic domain of a5 integrin subunit, and immunoprecipitation of VLA-5 integrin from cells known to express this integrin. Cell lines tested to express alpha5beta1, include: P19 cells, mouse melanoma B16F1 cells, mouse squamous carcoma SCC cells and thymocytes. Characterization by immunoprecipitation was typically analyzed by SDS-PAGE using 6% gels under non-reducing conditions.

Immunogen

The antibody was prepared by fusion of spleen cells from a Fisher rat immunized with mouse embryonal carcinoma P19 cells with myeloma P3X63Ag9.653 cells.

Application

Inhibition of cell adhesion to fibronectin: 10 μg/mL

Note that where cell adhesion to fibronectin involving other fibronectin receptors, such as VLA-4 (alpha4beta1) integrin, is involved, inhibition of cell adhesion by BMB5 alone may not be complete.

Immunoprecipitation from lysate of 10E6 cell equivalent: 2 μg

Flow cytometry: 10μg/mL

Western blot (non-denaturing gel)

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Integrin α5β1 Antibody, clone BMB5 is validated for use in FC, IP, WB, IH, FUNC for the detection of Integrin α5β1.

Physical form

Liquid, containing no preservatives.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Guadalupe Salgado-Lora et al.
Pathogens (Basel, Switzerland), 9(7) (2020-07-02)
Changes in the levels of reproductive hormones compromise the bovine innate immune response (IIR). Changes in 17β-estradiol (E2) and prolactin (bPRL) levels affect the IIR of bovine mammary epithelial cells (bMECs), the target tissue of these hormones. In this work
Decipher the dynamic coordination between enzymatic activity and structural modulation at focal adhesions in living cells.
Lu, S; Seong, J; Wang, Y; Chang, SC; Eichorst, JP; Ouyang, M; Li, JY; Chien, S; Wang, Y
Scientific Reports null
Ivan Medina-Estrada et al.
Mediators of inflammation, 2016, 6120509-6120509 (2016-04-02)
17β-Estradiol (E2), the predominant sexual hormone in females, is associated with the modulation of the innate immune response (IIR), and changes in its levels at parturition are related to intramammary infections, such as mastitis. In bovine mammary epithelial cells (bMECs)
Nayeli Alva-Murillo et al.
Frontiers in cellular and infection microbiology, 7, 78-78 (2017-04-01)
Bovine mammary epithelial cells (bMECs) contribute to mammary gland defense against invading pathogens, such as Staphylococcus aureus (intracellular facultative), which is recognized by TLR2. In a previous report, we showed that sodium octanoate [NaO, a medium chain fatty acid (C8)]
Yuefei Lou et al.
Reproductive biology and endocrinology : RB&E, 21(1), 85-85 (2023-09-16)
Adhesion of the implanting blastocyst involves the interaction between integrin proteins expressed by trophoblast cells and components present in the basement membrane of the endometrial luminal epithelium. Although several factors regulating integrins and their adhesion to fibronectin are already known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service