Skip to Content
MilliporeSigma
All Photos(1)

Documents

IPVH09120

Millipore

Immobilon® -P PVDF Membrane

10 sheets, 9 cm x 12 cm, 0.45 µm pore size, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22

product name

Immobilon®-P PVDF Membrane, 10 sheets, 9 cm x 12 cm, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

hydrophobic

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

filter L × W

9 cm × 12 cm

pore size

0.45 μm pore size

capacity

160 μg/cm2 adsorption capacity (insulin)
215 μg/cm2 adsorption capacity (BSA)
294 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with Coomassie brilliant blue
for use with India ink
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby
for use with Toluidine blue
for use with Transillumination

detection method

chemiluminescent
colorimetric
fluorometric
radioactive

shipped in

ambient

General description

Filter Code: IPVH
Primary Binding Mechanisms: Electrostatic, hydrophobic

Application

10 sheets, 9 cm x 12 cm, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.

Linkage

Replaces: INCP09120

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saumya Bhagat et al.
Blood cells, molecules & diseases, 84, 102459-102459 (2020-06-20)
Sterile Inflammation (SI), a condition where damage associated molecular patterns (DAMPs) released from dying cells, leads to TLR (Toll-like receptor) activation and triggers hypoxemia in circulation leading to venous thrombosis (VT) through tissue factor (TF) activation, but its importance under
Yu Li et al.
Oncology letters, 19(1), 323-333 (2020-01-04)
Non-small cell lung cancer (NSCLC) has long been one of the most lethal types of cancer due to its lack of typical clinical symptoms at early stages and high risk of tumour recurrence, even following complete surgical resection. Multicourse chemotherapy
Julie Gavard et al.
Molecular and cellular biology, 29(9), 2469-2480 (2009-03-04)
Most proangiogenic polypeptide growth factors and chemokines enhance vascular permeability, including vascular endothelial growth factor (VEGF), the main target for anti-angiogenic-based therapies, and interleukin-8 (IL-8), a potent proinflammatory mediator. Here, we show that in endothelial cells IL-8 initiates a signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service