Skip to Content
MilliporeSigma
All Photos(2)

Documents

HTMP1MAG-54K

Millipore

MILLIPLEX® Human TIMP Magnetic Bead Panel 1 - Immunology Multiplex Assay

TIMP Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple TIMP biomarkers in human serum, plasma and cell culture samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 92-101%
standard curve range: 20-20,000 pg/mL
(TIMP-1)

standard curve range: 49-50,000 pg/mL
(TIMP-2)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Tissue Inhibitors of Metalloproteinases (TIMPs) comprise a family of four inhibitors known as TIMP-1, TIMP-2, TIMP-3, and TIMP-4. Each member of the TIMP family has its own distinct profile of metalloproteinase inhibition. The matrix metalloproteinases (MMPs) are a family of zinc proteases involved in the breakdown of extracellular matrix (ECM) in normal physiological processes, such as embryonic development, tissue and bone remodeling, wound healing, tissue morphogenesis, angiogenesis, and tumor metastasis. MMPs are also known to be involved in the cleavage of cell surface receptors, the release of apoptotic ligands (such as the FAS ligand), cell proliferation, differentiation, and chemokine in/activation. The normal function of MMPs must be precisely regulated by their endogenous protein inhibitors, TIMPs . TIMPs inhibit MMPs by forming 1:1, non-covalent complexes with MMPs, thereby blocking access of substrates to the MMP catalytic site. Except for inhibition of active MMPs, TIMPs also exhibit several other biochemical and physiological/biological functions such as proMMP activation, cell growth promotion, matrix binding, inhibition of angiogenesis and the induction of apoptosis. Disruption of the MMP/TIMP balance can result in serious diseases such as arthritis, cardiovascular disorders, tumor growth and metastasis.

The MILLIPLEX® Human TIMP Panel 1 Bead kit (TIMP-1, -2) is to be used for serum/plasma samples.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
No significant cross-reactivity was observed among analytes in this panel.

Application

  • Analytes: TIMP-1, TIMP-2
  • Recommended Sample type: Serum or plasma
  • Recommended Sample dilution: 1:50
  • Assay Run Time: One day or overnight
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual analytes.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuuki Sasaki et al.
Biomolecules, 11(10) (2021-10-24)
This study aimed to explore whether cerebrospinal fluid (CSF) levels of matrix metalloproteinases (MMPs), and their inhibitors (TIMPs) were associated with brain amyloid deposition, cortical glucose metabolism, and white matter lesions (WMLs) in individuals with amnestic mild cognitive impairment (MCI).
Ulrike Müller et al.
Mediators of inflammation, 2015, 626530-626530 (2015-07-18)
In cystic fibrosis (CF) the upper (UAW) and lower airways (LAW) are reservoirs for pathogens like Pseudomonas aeruginosa. The consecutive hosts' release of proteolytic enzymes contributes to inflammation and progressive pulmonary destruction. Objectives were to assess dynamics of protease :
Johannes F Fahrmann et al.
Gastroenterology, 160(4), 1373-1383 (2020-12-18)
There is substantial interest in liquid biopsy approaches for cancer early detection among subjects at risk, using multi-marker panels. CA19-9 is an established circulating biomarker for pancreatic cancer; however, its relevance for pancreatic cancer early detection or for monitoring subjects
Yingqian Zhang et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 37(8), 2963-2974 (2016-11-26)
Blood-brain barrier (BBB) disruption plays an important role in pathophysiological progress of ischemic stroke. However, our knowledge of the dynamic change of BBB permeability and its mechanism remains limited. In the current study, we used a non-human primate (NHP) MCAO

Related Content

Immunoassays in cosmetics research assess product safety and effects on inflammation, aging, and tissue regeneration.

Immunoassays in cosmetics research assess product safety and effects on inflammation, aging, and tissue regeneration.

Immunoassays in cosmetics research assess product safety and effects on inflammation, aging, and tissue regeneration.

Immunoassays in cosmetics research assess product safety and effects on inflammation, aging, and tissue regeneration.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service