Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HMMP2MAG-55K

Millipore

MILLIPLEX® Human MMP Magnetic Bead Panel 2 - Immunology Multiplex Assay

Matrix Metalloproteinase Bead-Based Multiplex Assays using the Luminex technology enable the simultaneous analysis of multiple MMPs biomarkers in human serum, plasma and cell culture samples.

Synonym(s):

Human matrix metalloproteinase multiplex kit, Luminex® Human MMP immunoassay, Millipore human MMP panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 72-98%
standard curve range: 14-10,000 pg/mL
(MMP-9)

standard curve range: 27-20,000 pg/mL
(MMP-1)

standard curve range: 27-20,000 pg/mL
(MMP-10)

standard curve range: 548-400,000 pg/mL
(MMP-7)

standard curve range: 68-50,000 pg/mL
(MMP-2)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

MMPs (matrix metalloproteinases), a family of zinc proteases responsible for the breakdown of extracellular matrix (ECM), play a key role in normal physiological processes, such as embryonic development and tissue morphogenesis, tissue and bone remodeling, wound healing, and angiogenesis. These processes rely on MMPs′ role in the cleavage of cell surface receptors, the release of apoptotic ligands, cell proliferation and differentiation, and chemokine activity modulation. Similar in structure, MMPs are synthesized and secreted as inactive pro-enzymes that require proteolytic cleavage for activation. This process can be mediated by serine proteases or other MMPs. An increase in MMP expression occurs in response to a wide range of stimuli, including adhesion molecules, growth factors, cytokines, and hormones. Regulation of MMP activity is controlled primarily by TIMPs (tissue inhibitors of metalloproteinases). Therefore, disruption of the MMP/TIMP balance can result in arthritis, cardiovascular disease and tumor growth and metastasis.
MMP/TIMP research plays a significant role in achieving a deeper understanding of disease states such as chronic inflammation, cardiovascular disease, and cancer.

The MILLIPLEX® Human MMP Bead Panel 2 (MMP-1, -2, -7, -9, -10) will enable you to explore the modulation of and the function of MMP expression in multiple therapeutic areas.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
No significant cross-reactivity was observed among analytes in this panel.

Application

  • Analytes: MMP-1, MMP-2, MMP-7, MMP-9, MMP-10
  • Recommended Sample type: human serum, plasma, tissue/cell lystate and culture supernatant samples
  • Recommended Sample dilution: 1:20
  • Assay Run Time: One day
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: See protocol for sensitivities of individual analytes.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mittal Jasoliya et al.
Brain sciences, 10(6) (2020-06-14)
Recent advances in neurobiology have provided several molecular entrees for targeted treatments for Fragile X syndrome (FXS). However, the efficacy of these treatments has been demonstrated mainly in animal models and has not been consistently predictive of targeted drugs' response
Yu Yue et al.
BioMed research international, 2020, 2961742-2961742 (2020-07-01)
The present study investigated the profiles of matrix metalloproteinases (MMPs) and tissue inhibitors of metalloproteinases (TIMPs) of the aqueous humor (AH) and plasma (PL) in myopia patients, to determine whether there was an association between these levels with their axial
Livia V Souza et al.
Nutrition, metabolism, and cardiovascular diseases : NMCD, 30(1), 60-70 (2019-11-23)
The purpose of this study was to investigate whether an intervention with physical activity (PA) would promote positive effects on the angiogenic factors, mobilization, and functionality of circulating endothelial progenitor cells (EPCs) in children with low birth weight (LBW). Thirty-five
Ulrike Müller et al.
Mediators of inflammation, 2015, 626530-626530 (2015-07-18)
In cystic fibrosis (CF) the upper (UAW) and lower airways (LAW) are reservoirs for pathogens like Pseudomonas aeruginosa. The consecutive hosts' release of proteolytic enzymes contributes to inflammation and progressive pulmonary destruction. Objectives were to assess dynamics of protease :
Abstracts from the 43rd American Society of Andrology Annual Meeting, 21-24 April 2018, Portland, Oregon.
Andrology, 6 Suppl 1, 1-98 (2018-03-28)

Related Content

Simplify protein biomarker discovery with the MILLIPLEX® PLEXpedition 115-Plex Multiplex Screening Panel. Effortlessly screen cytokines and other markers with Luminex® technology.

Simplify protein biomarker discovery with the MILLIPLEX® PLEXpedition 115-Plex Multiplex Screening Panel. Effortlessly screen cytokines and other markers with Luminex® technology.

Simplify protein biomarker discovery with the MILLIPLEX® PLEXpedition 115-Plex Multiplex Screening Panel. Effortlessly screen cytokines and other markers with Luminex® technology.

Cancer biomarker detection through MILLIPLEX® multiplex cancer assays can accelerate studies of tumorigenic processes helping advance cancer research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service