MilliporeSigma
  • Changes of Proteases, Antiproteases, and Pathogens in Cystic Fibrosis Patients' Upper and Lower Airways after IV-Antibiotic Therapy.
Changes of Proteases, Antiproteases, and Pathogens in Cystic Fibrosis Patients' Upper and Lower Airways after IV-Antibiotic Therapy.
Mediators of inflammation (2015-07-18)
Ulrike Müller, Julia Hentschel, Wibke K Janhsen, Kerstin Hünniger, Uta-Christina Hipler, Jürgen Sonnemann, Wolfgang Pfister, Klas Böer, Thomas Lehmann, Jochen G Mainz
ABSTRACT
In cystic fibrosis (CF) the upper (UAW) and lower airways (LAW) are reservoirs for pathogens like Pseudomonas aeruginosa. The consecutive hosts' release of proteolytic enzymes contributes to inflammation and progressive pulmonary destruction. Objectives were to assess dynamics of protease : antiprotease ratios and pathogens in CF-UAW and LAW sampled by nasal lavage (NL) and sputum before and after intravenous- (IV-) antibiotic therapy. From 19 IV-antibiotic courses of 17 CF patients NL (10 mL/nostril) and sputum were collected before and after treatment. Microbiological colonization and concentrations of NE/SLPI/CTSS (ELISA) and MMP-9/TIMP-1 (multiplex bead array) were determined. Additionally, changes of sinonasal symptoms were assessed (SNOT-20). IV-antibiotic treatment had more pronounced effects on inflammatory markers in LAW, whereas trends to decrease were also found in UAW. Ratios of MMP-9/TIMP-1 were higher in sputum, and ratios of NE/SLPI were higher in NL. Remarkably, NE/SLPI ratio was 10-fold higher in NL compared to healthy controls. SNOT-20 scores decreased significantly during therapy (P = 0.001). For the first time, changes in microbiological patterns in UAW and LAW after IV-antibiotic treatments were assessed, together with changes of protease/antiprotease imbalances. Delayed responses of proteases and antiproteases to IV-antibiotic therapy were found in UAW compared to LAW.
Product Description
Millipore
MILLIPLEX® Human TIMP Magnetic Bead Panel 1 - Immunology Multiplex Assay, TIMP Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple TIMP biomarkers in human serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human MMP Magnetic Bead Panel 2 - Immunology Multiplex Assay, Matrix Metalloproteinase Bead-Based Multiplex Assays using the Luminex technology enable the simultaneous analysis of multiple MMPs biomarkers in human serum, plasma and cell culture samples.