Skip to Content
MilliporeSigma
All Photos(2)

Documents

MCYTMAG-70K-PX32

Millipore

MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Premixed 32 Plex - Immunology Multiplex Assay

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.

Synonym(s):

Luminex® Mouse cytokine immunoassay, Millipore Mouse cytokine immunoassay, Mouse cytokine Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

accuracy: 85-107%
standard curve range: 3.2-10,000 pg/mL

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The immune system is an organized complex network of biological structures and processes that protect against disease. For example, cytokine and chemokines mediate interactions between cells directly, regulating target immune cell responses. These responses result in inflammation where the immune system attempts to eradicate foreign antigens and begin the healing process. Consequently, the inflammatory process plays a key protective role in immunity. In addition, cytokine and chemokine research is essential in understanding the immune system and its multi-faceted response to most antigens, as well as disease states such as autoimmune disease, allergic reactions, sepsis, and cancer.

To identify specific cytokines involved in any inflammatory or immune response, it might be necessary to screen panels of cytokines, often requiring some level of automation and/or high throughput. Beads can make the process of automation and high throughput screening easier with features such as walk-away washing. Advantages even outside automation include:

More flexible plate and plate washer options
  • Improved performance with turbid serum/plasma samples
  • Assay results equivalent to non- beads
  • Automated washing avoids many problems associated with vacuum filtration washing


MILLIPLEX® Mouse Cytokine / Chemokine panel enables you to focus on the therapeutic potential of cytokines as well as the modulation of cytokine expression. Coupled with the Luminex® xMAP® platform in a bead format, you receive the advantage of ideal speed and sensitivity, allowing quantitative multiplex detection of dozens of analytes simultaneously, which can dramatically improve productivity.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: Eotaxin/CCL11, G-CSF, GM-CSF, IFN-γ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-17, IP-10, KC, LIF, LIX, MCP-1, M-CSF, MIG, MIP-1α, MIP-1β, MIP-2, RANTES, TNF-α, VEGF
  • Recommended Sample type: plasma, serum, or cell culture supernatant
  • Recommended Sample dilution: 1:2 diluted plasma or serum
  • Assay Run Time: Overnight or one day. For best results an overnight incubation is recommended.
  • Research Category: Inflammation & Immunology

Packaging

96 well plate
Premixed beads for your convenience.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities for individual cytokines.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulises Urzúa et al.
Biomolecules, 10(1) (2020-01-16)
In middle-aged women, the decline of ovarian follicle reserve below a critical threshold marks menopause, leading to hormonal, inflammatory, and metabolic changes linked to disease. The highest incidence and mortality of sporadic ovarian cancer (OC) occur at post-menopause, while OC
Jennifer L Johnson et al.
Neuron, 104(4), 665-679 (2019-10-06)
In humans, disruption of nonsense-mediated decay (NMD) has been associated with neurodevelopmental disorders (NDDs) such as autism spectrum disorder and intellectual disability. However, the mechanism by which deficient NMD leads to neurodevelopmental dysfunction remains unknown, preventing development of targeted therapies.
Montasir Elahi et al.
Journal of Alzheimer's disease : JAD, 51(4), 1209-1224 (2016-03-01)
Recent epidemiological evidence suggests that diabetes mellitus (DM) is a risk factor for Alzheimer's disease (AD). One of the pathological hallmarks of AD is hyperphosphorylated tau protein, which forms neurofibrillary tangles. Oxidative stress and the activation of inflammatory pathways are
Manju Kumari et al.
The Journal of clinical investigation, 126(8), 2839-2854 (2016-07-12)
The chronic inflammatory state that accompanies obesity is a major contributor to insulin resistance and other dysfunctional adaptations in adipose tissue. Cellular and secreted factors promote the inflammatory milieu of obesity, but the transcriptional pathways that drive these processes are
Anja Fuchs et al.
Journal of immunology (Baltimore, Md. : 1950), 202(10), 3020-3032 (2019-04-17)
The inflammatory response to infection or injury dramatically increases the hematopoietic demand on the bone marrow to replace effector leukocytes consumed in the inflammatory response. In the setting of infection, pathogen-associated molecular patterns induce emergency hematopoiesis, activating hematopoietic stem and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service