Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HCYTPAB

Millipore

MILLIPLEX® Human Cytokine Panel A + B Combo Pack

Synonym(s):

Human cytokine multiplex kit, Humaninterferon multiplex kit, Luminex human interferon panel, Luminexhuman cytokine immunoassay, Milliporehuman cytokine panel, humaninterferon panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

species reactivity

human

parameter

25 μL sample volume (per well)

technique(s)

multiplexing: suitable

assay range

inter-assay cv: ≤15%
intra-assay cv: ≤20%

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

Cytokines, chemokines, and growth factors are key mediators of immune system functions capable of signaling through autocrine, paracrine, and endocrine mechanisms. Their pleiotropic immunomodulatory properties allow these biomolecules to react to diverse stimuli and regulate the immune response, either by promoting or inhibiting inflammation. Growing research continues to highlight the role of the immune system in every facet of human health and disease.

Research category: Inflammation & Immunology
Research sub-category: Inflammation & Autoimmune Mechanisms; Inflammation

Application

Included in the 76-Plex Combo Pack: The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A Magnetic Bead 38-Plex Premixed Panel is a fixed 38-plex kit to be used for the simultaneous quantification of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: EGF;Eotaxin;G-CSF;GM-CSF;IFNα2;IFNγ;IL-1α;IL-1β;IL-1ra;IL-2;IL-3;IL-4;IL-5;IL-6;IL-7;IL-8;IL-10;IL-12 (p40);IL-12 (p70);IL-13;IL-15;IL-17A;IL-17E/IL-25;IL-17F;IL-18;IL-22;IP-10;MCP-1;M-CSF;MIG;MIP-1α;MIP-1β;PDGF-AA;PDGF-AB/BB;RANTES;TNFα;TNFβ;VEGF-A. RANTES cannot be plexed with other cytokines in this panel due to a required 1:100 dilution of plasma/serum samples.This premixed panel comes with a RANTES bead, so please plan your samples accordingly.

The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel B Magnetic Bead 38-Plex Premixed Panel is a fixed 38-plex kit to be used for the simultaneous quantification of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: 6Ckine, sCD137, sFAS, sFASL, APRIL, BAFF, BCA-1, CTACK, ENA-78, Eotaxin-2, Eotaxin-3, Granzyme A, Granzyme B, HMGB1, I-309, IFNβ, IFNω, IL-16, IL-20, IL-21, IL-23, IL-28A, IL-29, IL-31, IL-33, I-TAC, LIF, MCP-2, MCP-4, MIP-1δ, MIP-3α, Perforin, SCF, SDF-1, TARC, TPO, TRAIL, TSLP.

Included in the 96-plex Combo Pack: The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A is a 48-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: sCD40L;EGF;Eotaxin;FGF-2;Flt-3 ligand;Fractalkine;G-CSF;GM-CSF;GROα;IFNα2;IFNγ;IL-1α;IL-1β;IL-1ra;IL-2;IL-3;IL-4;IL-5;IL-6;IL-7;IL-8;IL-9;IL-10;IL-12 (p40);IL-12 (p70);IL-13;IL-15;IL-17A;IL-17E/IL-25;IL-17F;IL-18;IL-22;IL-27;IP-10;MCP-1;MCP-3;M-CSF;MDC (CCL22);MIG;MIP-1α;MIP-1β;PDGF-AA;PDGF-AB/BB;RANTES;TGFα;TNFα;TNFβ;VEGF-A. RANTES cannot be plexed with other cytokines in this panel due to a required 1:100 dilution of plasma/serum samples.This premixed panel comes with a RANTES bead, so please plan your samples accordingly.

The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel B Magnetic Bead Panel is a 48-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: 6Ckine, sCD137, sFAS, sFASL, APRIL, BAFF, BCA-1, CCL28, CTACK, CXCL16, ENA-78, Eotaxin-2, Eotaxin-3, GCP-2, Granzyme A, Granzyme B, HMGB1, I-309, IFNβ, IFNω, IL-11, IL-16, IL-20, IL-21, IL-23, IL-24, IL-28A, IL-29, IL-31, IL-33, IL-34, IL-35, I-TAC, LIF, Lymphotactin, MCP-2, MCP-4, MIP-1δ, MIP-3α, MIP-3β, MPIF-1, Perforin, SCF, SDF-1, TARC, TPO, TRAIL, TSLP.

Features and Benefits

Configuration: Premixed

Packaging

Two boxes; each to be analyzed separately.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panels A and B offer customizable options for analyzing nearly 100 cytokines.

MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panels A and B offer customizable options for analyzing nearly 100 cytokines.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service