Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EX0285

Supelco

Ethanol (denatured)

anhydrous

Synonym(s):

Alcohol, Denatured

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O
CAS Number:
Molecular Weight:
46.07
MDL number:
UNSPSC Code:
12352104

Product Name

Ethyl Alcohol, anhydrous, denatured

Quality Level

form

liquid

composition

ethanol, 92-94%
ethyl acetate, 1-2%
methanol, 3-5%

impurities

≤0.005% Acidity (as acetic acid)
≤0.5% Water (H2O)

color

APHA: ≤20

transition temp

flash point 11 °C

shipped in

ambient

storage temp.

room temp

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

anhydrous, denatured

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Physical form

100 volumes SD Alcohol #1 (1 volume Ethyl Acetate, 1 volume Hydrocarbon solvent, 1 volume MIBK)

Analysis Note

Acidity (as acetic acid)0.005% maxAssay (Ethanol)92-94%Assay (Ethyl acetate)1-2%Assay (Methanol)3-5%Color (APHA)20 maxFormClear liquidInfrared SpectrumConforms to standardNonvolatile residue0.0025% maxSpecific Gravity at 20/20°C0.789-0.793Water (H2O)0.5% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service