Skip to Content
MilliporeSigma
All Photos(1)

Documents

EX0282

Supelco

Ethanol (denatured)

≥95.0%, APHA: ≤20

Synonym(s):

Alcohol, Ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O
CAS Number:
Molecular Weight:
46.07
MDL number:
UNSPSC Code:
12352104

product name

Ethyl Alcohol, 95% denatured, special

Quality Level

Assay

95%

form

liquid

composition

ethanol, 84.0-86.0%
ethyl acetate, 4-5%
methanol, 3-4%
water, 7-10%

color

APHA: ≤20

transition temp

flash point 11.7 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

EX0282-8 is packaged in poly coated glass bottles

Analysis Note

Assay (Ethanol): 84.0-86.0%
Assay (Ethyl acetate): 4-5%
Assay (Methanol): 3-4%
Assay (Water): 7-10%
Color (APHA): 20 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.1 °F

Flash Point(C)

11.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service