Skip to Content
MilliporeSigma

Skip To

EX0280

Ethanol (denatured)

≥95.0%, APHA: ≤10

Synonym(s):

Alcohol, Ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C2H6O
CAS Number:
Molecular Weight:
46.07
UNSPSC Code:
12352104
MDL number:
Assay:
95%
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ethyl Alcohol, 95% denatured

Quality Segment

assay

95%

form

liquid

composition

ethanol, 86.0-89.0% , ethyl acetate, 1-2% , hydrocarbon solvent, as denaturant-1% , methanol, 3.0-5.0% , methyl isobutyl ketone, as denaturant-1% , water, 6-8%

impurities

≤0.005% Acidity (as acetic acid)

color

APHA: ≤10

transition temp

flash point 13.9 °C

shipped in

ambient

storage temp.

room temp

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

General description

EMPARTA solvents are designed for routine chemical synthesis, drying, purification, and critical labware cleaning in quality control environments. Engineered for precision, they deliver consistently reproducible results essential for rigorous validation and documentation.

Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.

Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.

Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.

Application

95% denatured

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Physical form

100 volumes SD Alcohol #1 (100 volumes Ethanol, 5 volumes Methanol)

Analysis Note

Acidity (as acetic acid)0.005% maxAssay (Ethanol)86.0-89.0%Assay (Ethyl acetate)1-2%Assay (Methanol)3-5%Assay (Water)6-8%Color (APHA)10 maxFormClear liquidInfrared SpectrumConforms to standardNonvolatile residue0.0025% maxSpecific Gravity at 20/20°C0.809-0.813

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EX0285EX0282EX0278
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

assay

95%

assay

-

assay

95%

assay

-

form

liquid

form

liquid

form

liquid

form

liquid

impurities

≤0.005% Acidity (as acetic acid)

impurities

≤0.005% Acidity (as acetic acid), ≤0.5% Water (H2O)

impurities

-

impurities

≤0.5% Water, ≤5 ng/L Capillary ECD responsive substances (as PCNB)

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

color

APHA: ≤10

color

APHA: ≤20

color

APHA: ≤20

color

APHA: ≤10


Still not finding the right product?

Explore all of our products under Ethyl Alcohol


signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

57.0 °F

flash_point_c

13.9 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters