Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

187380

Sigma-Aldrich

Ethyl alcohol, denatured

reagent grade

Synonym(s):

Ethanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
15101802
PubChem Substance ID:
grade:
denatured
reagent grade
Pricing and availability is not currently available.

grade

denatured
reagent grade

Quality Level

form

liquid

contains

1% ethyl acetate as denaturant
1% hydrocarbons as denaturant
1% methyl isobutyl ketone as denaturant
5% methanol as denaturant

impurities

≤0.15% water

General description

Ethanol (denatured), a primary alcohol, is a constituent of hairspray, disinfectants and cosmetics.[1] It can be synthesized either from molasses by fermentation or by hydration of ethylene.[2] It is made non-consumable by the addition of denaturants.[1]
The product is denatured ethyl alcohol and it contains following as denaturant:
  • 1% ethyl acetate
  • 1% hydrocarbons
  • 1% methyl isobutyl ketone
  • 5% methanol
Ethanol, a primary alcohol is an important raw material for the synthesis of various compounds. It can be synthesized either from molasses by fermentation or by hydration of ethylene. It acts as a hydrogen source and reaction medium during the transfer hydrogenation of carbonyl compounds in the presence of sodium hydroxide.[3] The impact of ethanol/water fumigation on the performance of diesel engines has been studied.[4] Denatured alcohol is also referred to as industrial methylated spirit.[5]

Application

Ethyl alcohol (denatured ethanol) may be used to compose the Nital etchant for use in microstructural analysis.[6]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Voltammetric Analysis on the Formation of Fe (OH)2 and FeCO3, and on the Reactivity of Passivation of Steel in Carbonate Solutions.
Eliyan FF, et al.
Journal of Materials Engineering and Performance, 24(6), 2473-2480 (2015)
Spencer LT, et al.
Tissue processing 6." Bancroft's Theory and Practice of Histological Techniques, Expert Consult: Online and Print, 7: Bancroft's Theory and Practice of Histological Techniques, 107-107 (2013)
Selection of highly effective rectification regimes for ethyl alcohol.
Vyaz'mina NA and Baranov DA.
Chemical and Petroleum Engineering, 42(9), 494-502 (2006)
Surrogate alcohol: what do we know and where do we go?
Lachenmeier DW, et al.
Alcoholism, Clinical and Experimental Research, 31(10), 1613-1624 (2007)
Sodium hydroxide-catalyzed transfer hydrogenation of carbonyl compounds and nitroarenes using ethanol or isopropanol as both solvent and hydrogen donor.
Wang D, et al.
J. Mol. Catal. A: Chem., 400, 14-21 (2015)

Questions

  1. What is the concentration of ethyl alcohol?

    1 answer
    1. This product is a denatured ethanol that includes the following additives:
      1% ethyl acetate as denaturant
      1% hydrocarbons as denaturant
      1% methyl isobutyl ketone as a denaturant
      5% methanol as denaturant
      The maximum allowable water content is 0.15%.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service