Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE2897

Sigma-Aldrich

Anti-phospho GATA6 (Ser37)

from rabbit

Synonym(s):

GATA-binding factor 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology), porcine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pSer37)

Gene Information

human ... GATA6(2627)

General description

Transcription factor GATA-6 (UniProt: Q92908; also known as GATA-binding factor 6) is encoded by the GATA6 gene (Gene ID: 2627) in human. GATA6 is a zinc-finger transcription factor that belongs to the six-member family of transcription factors that bind to the consensus sequence (A/T)GATA(A/G). GATA1, GATA2, and GATA3 are mainly expressed in hematopoietic cell lineages, while GATA4, GATA5, and GATA6 are predominantly expressed in the heart, gonads, and endodermal-derived tissues. It has two zinc-finger repeats localized to amino acids 390-414 and 444-468, which mediate interaction with LIM and cysteine-rich domains protein 1 (LMCD1). GATA-6 is shown to be important in the endodermal differentiation of organ tissues and has been indicated in proper lung development by controlling the late differentiation stages of alveolar epithelium and aquaporin-5 promoter activation. It has also been linked to the production of LIF that encourages proliferation of endodermal embryonic stem cells and blocks early epiblast differentiation. GATA6 is also required for definitive endoderm specification in human embryonic stem cells and induced pluripotent stem cells. Phosphorylation of GATA6 by GSK-3 on serine 37 is reported to promote its degradation, thereby preventing GATA6 from repressing transcripts that are induced by TNF and attenuated by insulin. Two isoforms of GATA-6 have been reported that are produced by alternative initiation. (Ref.: Chitforoushzadeh, Z., et al. (2016). ScienceSignaling 9 (431), ra59; Tiyaboonchai, A., et al. (2017). Stem Cell Reports 8 (3); 589-604).

Specificity

This rabbit polyclonal antibody detects GATA6 phosphorylated on Serine 37.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 37 of human Transcription factor GATA-6.

Application

Anti-phospho GATA6 (Ser37), Cat. No. ABE2897, is a highly specific rabbit polyclonal antibody that targets Transcription factor GATA-6 (GATA-binding factor 6) and has been tested for use in Dot Blot, Immunoprecipitation, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:500 dilution from a representative lot detected phospho GATA6 (Ser37) in 293T cells transfected with 3XFLAG-tagged GATA6L.

Western Blotting Analysis: A representative lot detected phospho GATA6 (Ser37) in Western Blotting applications (Chitforoushzadeh, Z., et. al. (2016). Sci Signal. 9(431):ra59).

Immunoprecipitation Analysis: 4 µg/mL from a representative lot immunoprecipitated phospho GATA6 (Ser37) in 293T cells were transfected with 3xFLAG-GATA6 (Courtesy of Dr. Kevin A. Janes at University of Virginia).

Immunoprecipitation Analysis: A representative lot immunoprecipitated phospho GATA6 (Ser37) in Immunoprecipitation applications (Chitforoushzadeh, Z., et. al. (2016). Sci Signal. 9(431):ra59).

Quality

Dot Blot Analysis: A 1:500 dilution from a representative lot detected phospho GATA6 (Ser37) in Phosphorylated GATA6 peptide.

Target description

~75 kDa observed; 60.03 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service