Skip to Content
MilliporeSigma
All Photos(3)

Documents

MAB3422

Sigma-Aldrich

Anti-Histone Antibody, clone H11-4

clone H11-4, Chemicon®, from mouse

Synonym(s):

Anti-Histone antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

H11-4, monoclonal

species reactivity

hamster, bovine, opossum, rat, human, Xenopus

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H2AC21(317772)

Related Categories

General description

There are 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. The five major classes of histones are H1/H5, H2A, H2B, H3, and H4. Histones H2A, H2B, H3 and H4 are known as the core histones, while histones H1 and H5 are known as the linker histones. Histones feature a main globular domain and a long N terminal tail which are involved with the structure of the nucleosomes of the ′beads on a string′ structure. They are the chief protein components of chromatin, acting as spools around which DNA winds, and play a role in gene regulation.

Specificity

Due to the highly conserved nature of histones, this antibody is predicted to have wide species reactivity.
The antibody binds specifically to an antigen determinant, which is present on all five histone proteins. The antibody recognizes the histones H1, H2A, H2B, H3 and H4 with comparable sensitivity.

Immunogen

Purified nuclear fractions of HeLa cells

Application

Detect Histone with Anti-Histone Antibody, clone H11-4 (Mouse Monoclonal Antibody), that has been shown to work in ICC, IHC & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western Blot Analysis:
1:500 dilution of a previous lot was used.

Immunocytochemistry:
A 1-5 µg/mL concentration of a previous lot was used on Immunocytochemistry.

Immunohistochemistry:
A 1-5 μg/mL concentration of a previous lot was used on Immunohistochemistry.

Optimal working dilutions must be determined by end user.

Quality

Routinely evaluated by Western Blot on Jurkat lysates.

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG1 in buffer containing 0.02M phosphate buffer, pH 7.6 with 0.25M NaCl and 0.1% sodium azide

Storage and Stability

Stable for 6 months at 2-8ºC in undiluted aliquots from date of receipt.
The Iyophilized antibody is stable at 2–8°C. Store the reconstituted antibody in aliquots at -15 to -25°C for up to 12 months.Repeated freezing and thawing should be avoided.

Analysis Note

Control
All tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenzhou Hong et al.
Journal of innate immunity, 1(3), 215-224 (2009-01-01)
Nontypeable Haemophilus influenzae (NTHi) is a leading cause of acute and chronic otitis media, which are a major public health problem worldwide. The persistence of NTHi during chronic and recurrent otitis media infections involves multicellular biofilm communities formed within the
Bing Pang et al.
Infection and immunity, 85(9) (2017-07-05)
Haemophilus parainfluenzae is a nutritionally fastidious, Gram-negative bacterium with an oropharyngeal/nasopharyngeal carriage niche that is associated with a range of opportunistic infections, including infectious endocarditis and otitis media (OM). These infections are often chronic/recurrent in nature and typically involve bacterial
Srimoyee Ghosh et al.
The Journal of biological chemistry, 291(25), 13014-13027 (2016-04-09)
Covalent modification of histones is a fundamental mechanism of regulated gene expression in eukaryotes, and interpretation of histone modifications is an essential feature of epigenetic control. Bromodomains are specialized binding modules that interact with acetylated histones, linking chromatin recognition to
Ershun Zhou et al.
Frontiers in immunology, 10, 1131-1131 (2019-06-14)
Given that B. besnoiti tachyzoites infect host endothelial cells of vessels in vivo, they become potential targets for professional phagocytes [e.g., polymorphonuclear neutrophils (PMN)] when in search for adequate host cells or in case of host cell lysis. It was
The transcriptional corepressor, PELP1, recruits HDAC2 and masks histones using two separate domains.
Choi, YB; Ko, JK; Shin, J
The Journal of Biological Chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service