Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC349

Sigma-Aldrich

Anti-ABCA7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

ATP-binding Cassette Sub-family A (ABC1) member 7, ABC1, ABC Transporter 1, ATP-binding Cassette 1, ATP-binding Cassette Transporter 1, CERP, ABCA-SSN, Autoantigen SS-N, Macrophage ABC transporter, ABCA7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ABCA7(10347)

General description

ATP-binding cassette sub-family A member 7 (ABCA7), also known as ABCA-SSN and Macrophage ABC transporter, is a full-size ABC transporter consisting of two sets of the multiple membrane-spanning domains plus the Walker motifs for the ATP interaction. It uses ATP hydrolysis to catalyze the transport of various molecules across cell membranes. ABCA7 is involved in phagocytosis by macrophages of apoptotic cells, binds to APOA1, may function in phospholipid and cholesterol efflux from cells, and may regulate cellular ceramide homeostasis during keratinocyte differentiation The high ABCA7 expression levels in peripheral leukocytes, thymus, spleen and bone marrow indicates a role in the immune system lipid homeostasis. ABCA1 is also highly expressed in fetal tissues, especially the liver.

Immunogen

KLH-conjugated linear peptide corresponding to human ABCA7.

Application

Anti-ABCA7 Antibody is a highly specific rabbit polyclonal antibody, that targets ABCA7 & has been tested in western blotting & ICC.
Immunocytochemistry Analysis: 5 µg/mL from a representative lot detected ABCA7 in HEK293 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1µg/mL of this antibody detected ABCA7 in 15 µg of HEK293 cell lysate.

Target description

~234 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 2 isoforms at ~219 kDa and ~234 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service