Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABN465

Sigma-Aldrich

Anti-PICALM Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Phosphatidylinositol-binding clathrin assembly protein, LAP, CALM, CLTH, Clathrin assembly lymphoid myeloid leukemia protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PICALM(8301)

General description

Phosphatidylinositol-binding clathrin assembly protein (PICALM), also known as the clathrin assembly lymphoid myeloid leukemia (CALM) protein, co-localizes with clathrin at clathrin-coated areas of the plasma membrane and in the Golgi area as a component of the coated pit internalization machinery. PICALM fusion proteins have been implicated in acute myeloid and T-cell acute lymphoblastic leukemia, and chromosomal aberrations are found in diffuse histiocytic lymphomas. PICALM is also expressed in neuronal synapses where it facilitates the endocytosis of VAMP2. PICALM variants are associated with Alzheimer′s disease and are thought to be involved in the endocytosis and recycling of the amyloid protein precursor (APP).

Specificity

This antibody can recognize all isoforms of PICALM.

Immunogen

KLH-conjugated linear peptide corresponding to human PICALM.

Application

Anti-PICALM Antibody is a highly specific rabbit polyclonal antibody, that targets Phosphatidylinositol-binding clathrin assembly protein & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in EL4 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected PICALM in 15 µg of EL4 cell lysate.

Target description

~62 kDa and ~71 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 5 isoforms between ~60 kDa and ~71 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
EL4 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service