Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB1765P

Sigma-Aldrich

Anti-Dopamine D1A Receptor Antibody, CT, cytoplasmic

Chemicon®, from rabbit

Synonym(s):

Anti-dadr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, pig

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DRD1(1812)

Specificity

Dopamine Receptor D1A. An antibody made to the D1R peptide has been shown to detect 2 proteins of 50 and 75 kDa in solubilized hippocampal membranes by Western blot. The 75 kDa band was not visible in striatum (Huang et al. 1992). Approximately 100-400 μg of protein/lane should be loaded to visualize the 75 kDa band. By immunohistochemistry D1R has been localized in various regions of the rat brain (Huang et al. 1992).

May recognize porcine or bovine (100 % similarity) or human (78% similarity).

Immunogen

A 13 amino acid peptide sequence (amino acids 403-415) from rat D1A (Monsma et al. 1990; Zhou et al. 1990; Zhou et al. 1992; O′Dowd et al. 1990). This peptide is predicted to be cytoplasmic.
Epitope: C-terminus, cytoplasmic

Application

Anti-Dopamine D1A Receptor Antibody, C-terminus, cytoplasmic is an antibody against Dopamine D1A Receptor for use in ELISA, IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1-10 μg/mL (Chemiluminescence technique). An antibody made to this peptide has been shown to detect 2 proteins of 50 and 75 kDa in solubilized hippocampal membranes. The 75 kDa band was not visible in striatum (Huang et al. 1992; O′Connel et al. 1995). Approximately 100-400 μg of protein/lane should be loaded to visualize the 75 kDa band (Huang et al. 1992; O′Connel et al. 1995) that is present in relatively small amounts.

Immunohistochemistry: 2-10 μg/mL on paraformaldehyde fixed sections.

ELISA: 0.5-1.0 μg/mL (1 μg/mL D1A peptide (Cat. Number AG259)/well)

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in phosphate buffered saline, pH 7.4, containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glenn M Dallérac et al.
Neuromolecular medicine, 18(1), 146-153 (2016-01-20)
Altered dopamine receptor labelling has been demonstrated in presymptomatic and symptomatic Huntington's disease (HD) gene carriers, indicating that alterations in dopaminergic signalling are an early event in HD. We have previously described early alterations in synaptic transmission and plasticity in
Violeta-Maria Caragea et al.
Brain structure & function, 229(3), 609-637 (2023-08-24)
The dopaminergic (DA) system regulates both motor function, and learning and memory. The cerebellum supports motor control and the acquisition of procedural memories, including goal-directed behavior, and is subjected to DA control. Its fastigial nucleus (FN) controls and interprets body
M Dean Graham et al.
The European journal of neuroscience, 42(12), 3138-3148 (2015-11-05)
Dopamine (DA) transmission in the medial preoptic area (mPOA) plays a critical role in the control of appetitive sexual behaviour in the female rat. We have shown previously that a DA D1 receptor (D1R)-mediated excitatory state appears to occur in
K L Conrad et al.
Neuroscience, 169(1), 182-194 (2010-05-04)
Dopamine receptors (DARs) in the nucleus accumbens (NAc) are critical for cocaine's actions but the nature of adaptations in DAR function after repeated cocaine exposure remains controversial. This may be due in part to the fact that different methods used
Xinquan Wang et al.
Free radical biology & medicine, 76, 242-250 (2014-09-23)
Adverse environment in early life can modulate the adult phenotype, including blood pressure. Lipopolysaccharide (LPS) exposure in utero results in increased blood pressure in the offspring, but the exact mechanisms are not clear. Studies have shown that the renal dopamine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service