Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB1784P

Sigma-Aldrich

Anti-Dopamine D1A Receptor Antibody, 4th Extracellular Domain

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DRD1(1812)

Specificity

Dopamine D1A Receptor (D1R). The rat D1A receptor is a 446 amino acid, G-protein coupled transmembrane receptor protein (Monsma, 1990; Zhou, 1990; Zhou, 1992; O′Dowd, 1990). The immunogen sequence has 88% sequence similarity with human and 77% with porcine.

Immunogen

A nine amino acid peptide corresponding to the 4th extracellular domain of the rat D1A receptor. The immunogen is absolutely conserved in the mouse D1A receptor and shows 88% homology with human and 77% with porcine and bovine D1A receptors. The peptide shows no significant sequence similarity with other dopamine receptors (D1B, D2-D5).
Epitope: 4th Extracellular Domain

Application

Detect Dopamine D1A Receptor using this Anti-Dopamine D1A Receptor Antibody, 4th Extracellular Domain validated for use in ELISA, IH & WB.
Immunohistochemistry: 2-10 μg/mL on paraformaldehyde fixed tissue sections. D1R has been localized in various regions of the rat brain (Huang, 1992; O′Connel, 1995)

Western blot: 1-10 μg/mL (Chemiluminescence technique). An antibody made to this peptide has been shown to detect 2 proteins of 50 and 75 kDa in solubilized hippocampal membranes. The 75 kDa band was not visible in striatum (Huang, 1992; O′Connel, 1995). Approximately 100-400 μg of protein/lane should be loaded to visualize the 75 kDa band (Huang, 1992; O′Connel, 1995) that is present in relatively small amounts.

ELISA: 0.5-1.0 μg/mL using 1 μg/mL of control peptide per well.

The immunogen peptide is available (Cat. No. AG266) for use in ELISA or antibody blocking studies. AG266 is not recommended for use as a western blot positive control.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.2% BSA and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months from date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: rat striatal membranes (western blot)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cátia Fernandes-Cerqueira et al.
BioMed research international, 2013, 397391-397391 (2013-08-21)
The edema formation in nephrotic syndrome (NS) is associated with a blunted response to atrial natriuretic peptide (ANP). The natriuretic effects of ANP have been related to renal dopamine D1-receptors (D1R). We examined the interaction between ANP and renal D1R
Glenn S Belinsky et al.
Stem cells and development, 22(10), 1522-1540 (2013-01-05)
We tested whether dopaminergic drugs can improve the protocol for in vitro differentiation of H9 human embryonic stem cells (hESCs) into dopaminergic neurons. The expression of 5 dopamine (DA) receptor subtypes (mRNA and protein) was analyzed at each protocol stage
Cecile Hryhorczuk et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 41(3), 811-821 (2015-07-15)
Overconsumption of dietary fat is increasingly linked with motivational and emotional impairments. Human and animal studies demonstrate associations between obesity and blunted reward function at the behavioral and neural level, but it is unclear to what degree such changes are
Tyler W Stradleigh et al.
Progress in retinal and eye research, 48, 181-202 (2015-04-22)
Immunohistochemical and ex vivo anatomical studies have provided many glimpses of the variety, distribution, and signaling components of vertebrate retinal neurons. The beauty of numerous images published to date, and the qualitative and quantitative information they provide, indicate that these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service