Skip to Content
MilliporeSigma

Skip To

MAB369

Anti-Dopamine Transporter Antibody, NT, clone DAT-Nt

culture supernatant, clone DAT-Nt, Chemicon®

Synonym(s):

DAT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
DAT-Nt, monoclonal
Application:
ICC, IHC, WB
Citations:
317
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

DAT-Nt, monoclonal

species reactivity

rat, human, mouse, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC6A3(6531)

General description

The transmembrane dopamine transporter (DAT) is located on the presynaptic nerve terminal and is responsible for terminating dopaminergic transmission by transporting dopamine from the synaptic cleft into the dopaminergic neuron (reuptake). Dopaminergic pathways have been strongly implicated in reward and addiction, motivation, alcoholism, ADHD and degenerative motor diseases such as Parkinson′s, Huntington′s and Chorea.
~ 70-85 kDa

Immunogen

Epitope: N-terminus
N-terminus of human dopamine transporter fused to Glutathione S-transferase.

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-Dopamine Transporter Antibody, N-terminus, clone DAT-Nt is validated for use in IC, IH, WB for the detection of Dopamine Transporter.
Western blot: Recognizes a diffuse band at approximately 70-85 kDa on western blots of extracts (20 ug total protein) from human caudate, putamen, and nucleus accumbens.

Immunohistochemistry: 4% paraformaldehyde fixed tissue (care should be taken not to over-fix tissue); perfusion followed by less than 90 minutes post-fixation, then cryoprotect. Suggested working dilution 1:1,000-1:10,000. If using on rat tissue, absorbed anti-rat secondary antibodies are recommended, and the use of rat PAP systems, or ABC detection will enhance sensitivity.

Immunocytochemistry: 1:5,000 to 1:10,000.

Immunoblotting (not recommended for use on rat)

Optimal working dilutions and protocols must be determined by end user.



IMMUNOHISTOCHEMISTRY PROTOCOL FOR MAB369

This antibody has been used successfully on 30 μm, free floating, 4% paraformaldehyde fixed mouse brain tissue. All steps are performed under constant agitation. Suggested protocol follows.

1) 3 x 10 minute washes in TBS (with or without 0.25% Triton).

2) Incubate for 30 minutes in TBS with 3% serum (same as host from secondary antibody).

3) Incubate primary antibody diluted appropriately in TBS with 1% serum (same as host from secondary antibody) (with or without 0.25% Triton) for 2 hours at room temperature followed by 16 hours at 4°C.

4) 3 x 10 minute washes in TBS.

5) Incubate with secondary antibody diluted appropriately in TBS with 1% serum (same as host from secondary antibody).

6) 3 x 10 minute washes in TBS.

7) ABC Elite (1:200 Vector Labs) in TBS.

8) 2 x 10 minute washes in TBS.

9) 1 x 10 minute wash in phosphate buffer (no saline).

10) DAB reaction with 0.06% NiCl added for intensification.

11) 2 x 10 minute washes in PBS.

12) 1 x 10 minute wash in phosphate buffer (no saline).

Biochem/physiol Actions

Recognizes Dopamine transporter, N-terminus. Shows no cross reactivity to the closely related serotonin and norepinephrine transporters (Miller, 1997). Immunolocalization of DAT on paraformaldehyde fixed frozen sections of human brain using MAB369 shows dense punctate staining throughout the caudate, putamen and accumbens (Miller, 1997).

Physical form

UnPurified tissue culture supernatant, filtered through a 0.2 μm membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL.
Unpurified

Preparation Note

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Control: Brain (caudate, putamen, and nucleus accumbens) tissue

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB1766AB1591PAB5802
description

culture supernatant, clone DAT-Nt, Chemicon®

description

Chemicon®, from rabbit

description

Chemicon®, from rabbit

description

Chemicon®, from rabbit

clone

DAT-Nt, monoclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

rat, human, mouse, monkey

species reactivity

monkey, human

species reactivity

rat, mouse

species reactivity

human, monkey

Gene Information

human ... SLC6A3(6531)

Gene Information

human ... SLC6A3(6531)

Gene Information

human ... SLC6A3(6531)

Gene Information

human ... SLC6A3(6531)

antibody form

culture supernatant

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

biological source

rat

biological source

rabbit

biological source

rabbit

biological source

rabbit


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters