Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.45501

Sigma-Aldrich

Thiourea

for photovoltaic

Synonym(s):

Thiourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4N2S
CAS Number:
Molecular Weight:
76.12
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-543-5
NACRES:
NA.22

Quality Level

form

solid

autoignition temp.

440 °C (Dust)

potency

1750 mg/kg LD50, oral (Rat)
>2800 mg/kg LD50, skin (Rabbit)

pH

6-8 (20 °C, 50 g/L in H2O)

mp

169-173 °C

solubility

137 g/L

density

1.405 g/cm3 at 20 °C

bulk density

640 kg/m3

storage temp.

no temp limit

SMILES string

S=C(N)N

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)

InChI key

UMGDCJDMYOKAJW-UHFFFAOYSA-N

Analysis Note

Assay: ≥ 98.0 %
Identity (IR): passes test
Appearance of solution (50 g/l; water): clear to nearly clear and colourless
Appearance (visual): white to off white crystalin powder
Melting point: 174 - 178 °C
Al (Aluminium): ≤ 5 ppm
As (Arsenic): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cr (Chromium): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 5 ppm
Mn (Manganese): ≤ 5 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Sb (Antimony): ≤ 5 ppm
Sn (Tin): ≤ 5 ppm
Zn (Zinc): ≤ 5 ppm
Sulfated ash (600 °C): ≤ 0.1 %
Loss on Drying (105°C): ≤ 0.5 %
Sensitivity to bismuth: passes test

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service