Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

560154

Sigma-Aldrich

1-(3-Carboxyphenyl)-2-thiourea

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(S)NHC6H4CO2H
CAS Number:
Molecular Weight:
196.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

187-191 °C (lit.)

functional group

amine
carboxylic acid
thiourea

SMILES string

NC(=S)Nc1cccc(c1)C(O)=O

InChI

1S/C8H8N2O2S/c9-8(13)10-6-3-1-2-5(4-6)7(11)12/h1-4H,(H,11,12)(H3,9,10,13)

InChI key

JRQJYVACDJEUDZ-UHFFFAOYSA-N

General description

1-(3-Carboxyphenyl)-2-thiourea (CPTU) is a thiourea derivative containing electron donor atoms (N and S). It acts as a ligand and readily form complexes with Hg(II).

Application

1-(3-Carboxyphenyl)-2-thiourea functionalized mesoporous silica SBA-15 (CPTU-SBA-15) has been used for the solid phase extraction of Hg(II).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of aqueous toxic Hg (II) by functionalized mesoporous silica materials.
Zhang L, et al.
Journal of Chemical Technology and Biotechnology, 87(10), 1473-1479 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service