Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1758

Supelco

Thiourea

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Thiourea, Sulfourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CSNH2
CAS Number:
Molecular Weight:
76.12
Beilstein:
605327
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

propylthiouracil

CofA

current certificate can be downloaded

packaging

pkg of 200 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

170-176 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

NC(N)=S

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)

InChI key

UMGDCJDMYOKAJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Thiourea is a chaotropic agent or strong denaturant. It has been used to solubilize membrane and organelle specific proteins for analysis by two-dimensional gel electrophoresis. Thiourea is a free radical scavenger of the peroxide radical. It was shown to inhibit lipid peroxidation and UV-induced crosslinking of collagen. As a chelator of cuprous copper, thiourea inhibits this intermediate in oxidative reactions of cupric ion. This process afforded protection of bovine albumin from copper-mediated oxidative reactions.

Application

Chaotropic agent; strong denaturant. Increases solubility and recovery of proteins
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8226 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service