Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.51080

Sigma-Aldrich

Bis-Boc-thiourea

Novabiochem®

Synonym(s):

Bis-Boc-thiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20N2O4S
CAS Number:
Molecular Weight:
276.35
MDL number:
UNSPSC Code:
12352108

Quality Level

Assay

≥98.0% (HPLC)

form

powder

manufacturer/tradename

Novabiochem®

storage temp.

2-8°C

SMILES string

S=C(NC(=O)OC(C)(C)C)NC(=O)OC(C)(C)C

InChI

1S/C11H20N2O4S/c1-10(2,3)16-8(14)12-7(18)13-9(15)17-11(4,5)6/h1-6H3,(H2,12,13,14,15,18)

InChI key

CSOJECDGWHHWRS-UHFFFAOYSA-N

General description

Reagent for the guanidation of amines.

Linkage

Replaces: 01-63-0135

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (DC(012A)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service