Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.07971

Sigma-Aldrich

Succinimide

for synthesis

Synonym(s):

Succinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NO2
CAS Number:
Molecular Weight:
99.09
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-635-6
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 50 °C)

Quality Level

Assay

≥98.0% (GC)

form

powder

potency

14000 mg/kg LD50, oral (Rat)

pH

4-6 (200 g/L in H2O)

mp

121-124 °C

transition temp

flash point 201 °C

solubility

330 g/L

density

1.4 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1C(=O)CCC1=O

InChI

1S/C4H5NO2/c6-3-1-2-4(7)5-3/h1-2H2,(H,5,6,7)

InChI key

KZNICNPSHKQLFF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 120 °C
Melting range (upper value): ≤ 126 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service