Skip to Content
MilliporeSigma
N-Chlorosuccinimide 98%

N-Chlorosuccinimide

N-Chlorosuccinimide 98%
Synonym(s):
N-Chlorosuccinimide
Empirical Formula (Hill Notation):
C4H4ClNO2
CAS Number:
Molecular Weight:
133.53

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing