Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.03463

Sigma-Aldrich

1-Decanol

for synthesis

Synonym(s):

1-Decanol, Capric alcohol, Decyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9OH
CAS Number:
Molecular Weight:
158.28
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-956-9
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

254 °C

potency

4720 mg/kg LD50, oral (Rat)
3560 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-5.7 % (v/v)

bp

220-235 °C/1013 hPa

mp

5-7 °C

transition temp

flash point 95 °C (DIN 51758)

density

0.8 g/cm3 at 20 °C (DIN 51757)

storage temp.

2-30°C

InChI

1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3

InChI key

MWKFXSUHUHTGQN-UHFFFAOYSA-N

Application

  • Enhancement of fuel properties: 1-Decanol is used as a cosolvent in diesel-methanol blends to improve combustion, emission, and stability characteristics, offering potential enhancements for biofuel applications (El-Seesy et al., 2022).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.829 - 0.830
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - Pensky-Martens closed cup

Flash Point(C)

95 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Summary application report for analysis of moisture in Transformer oil

Summary application report for analysis of moisture in Petroleum

Titration of hydrocarbons may require solubilizers for unpolar substances; recommend long-chain alcohols or chloroform.

Titration of hydrocarbons may require solubilizers for unpolar substances; recommend long-chain alcohols or chloroform.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service