Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.03462

Sigma-Aldrich

1-Dodecanol

for synthesis

Synonym(s):

1-Dodecanol, Dodecyl alcohol, Lauryl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OH
CAS Number:
Molecular Weight:
186.33
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-982-0
NACRES:
NA.22
Assay:
≥98% (GC)
bp:
229 °C
vapor pressure:
0.08 mbar ( 38 °C)
Pricing and availability is not currently available.

vapor pressure

0.08 mbar ( 38 °C)

Quality Level

Assay

≥98% (GC)

form

paste

autoignition temp.

255 °C

potency

>5000 mg/kg LD50, oral (Rat)

bp

229 °C

mp

24 °C

transition temp

flash point 134.8 °C

solubility

0.004 g/L

density

0.9 g/cm3 at 16 °C

storage temp.

2-30°C

SMILES string

OCCCCCCCCCCCC

InChI

1S/C12H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3

InChI key

LQZZUXJYWNFBMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Facile synthesis of microencapsulated 1-dodecanol/melamine-formaldehyde: This study discusses the development of phase change material for thermal energy storage, using 1-dodecanol encapsulated in melamine-formaldehyde. (Kumar et al., 2021).
  • PDMS modified membranes by 1-dodecanol: This research examines the effect of 1-dodecanol on ethanol removal via pervaporation, highlighting improvements in separation processes. (León and Fontalvo, 2019).
  • Liquid–liquid equilibria for 1-dodecanol system: The study presents experimental data and predictions on the liquid–liquid equilibria involving 1-dodecanol, valuable for chemical engineering applications. (Pérez et al., 2016).
  • Nanometer confinement induces nematic order in 1-Dodecanol: Investigating the phase behavior of 1-dodecanol under nanometer confinement, this research provides insights into material science and nanotechnology. (Yamaguchi et al., 2018).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 22 °C
Melting range (upper value): ≤ 26 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service