U1001
1-Undecanol
97%
Synonym(s):
Alcohol C11, Undecyl alcohol
About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.44 (lit.)
bp
146 °C/30 mmHg (lit.)
mp
11 °C (lit.)
density
0.83 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCO
InChI
1S/C11H24O/c1-2-3-4-5-6-7-8-9-10-11-12/h12H,2-11H2,1H3
InChI key
KJIOQYGWTQBHNH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
It can be also used as a solvent in homogeneous liquid-liquid microextraction method.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
226.4 °F - closed cup
Flash Point(C)
108 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service