Skip to Content
MilliporeSigma

8.08468

1-Undecanol

for synthesis

Synonym(s):

1-Undecanol, Undecyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)10OH
CAS Number:
Molecular Weight:
172.31
UNSPSC Code:
12352104
EC Index Number:
203-970-5
NACRES:
NA.22
MDL number:
Assay:
≥97.5% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<1 hPa ( 20 °C)

Quality Segment

assay

≥97.5% (GC)

form

liquid

potency

3000 mg/kg LD50, oral (Rat)

kinematic viscosity

20.03 cSt(20 °C)

mp

16 °C

transition temp

flash point 108 °C

density

0.83 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCCCCCCCCCC

InChI

1S/C11H24O/c1-2-3-4-5-6-7-8-9-10-11-12/h12H,2-11H2,1H3

InChI key

KJIOQYGWTQBHNH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.832 - 0.833
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


Still not finding the right product?

Explore all of our products under 1-Undecanol


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

226.4 °F - closed cup

flash_point_c

108 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library