Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

73070BC

Millipore

GDS Rotor-Gene Q Thermocycler

Advanced rotary thermocycler for use with Assurance® GDS PCR assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102421

Quality Level

manufacturer/tradename

Assurance GDS®
BioControl Systems 73070

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with BioControl GDS

storage temp.

room temp

General description

Assurance® GDS (Genetic Detection System) is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The unique centrifugal rotary design of the Rotor-Gene® Q makes it the most precise and versatile thermocycler currently available. Each tube spins in a chamber of moving air, keeping all samples at precisely the same temperature during rapid thermal cycling thereby enabling uniform detection. When each tube aligns with the detection optics, the sample is illuminated, and the fluorescent signal is rapidly collected from a single, short optical pathway. This thermal and optical uniformity results in sensitive, precise, and fast PCR analysis. It also eliminates sample-to-sample variations and edge effects. These are unavoidable in traditional block-based instruments due to temperature gradients across the block and multiple, complex optical pathways.

Application

GDS Rotor-Gene® Q Thermocycler is used with Assurance® GDS assays for the detection of pathogenic organisms in foods, ingredients, and environmental samples without the need for optimization of reaction and cycling conditions.

Features and Benefits

The rotary design delivers:
  • Tube-to-tube variation ±0.02°C for uniform detection
  • Pre-loaded firmware specific to Assurance® GDS assays
  • Negligible equilibration times for fast run-cycling
  • Complete confidence in your results

Components

  • GDS Rotor-Gene Q
  • User Manual
  • GDS Gel Cooling Block (pack of 2)

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
Rotor-Gene is a registered trademark of Qiagen GmbH

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service