Skip to Content
MilliporeSigma

71009BC

Assurance® GDS Listeria spp. Tq Kit

suitable for PCR testing in Food and Beverages applications

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106302
NACRES:
NA.85
Agency:
PCR, Immunomagnetic Separation (IMS), AOAC® (PTM), Health Canada, MicroVal
Suitability:
Listeria spp.
Manufacturer/tradename:
BioControl Systems 71009-100
Application(s):
food and beverages
pathogen testing
Technique(s):
PCR: suitable
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

GDS Listeria spp. Tq, BioControl, Molecular based PCR test for the detection of Listeria spp. in food and environmental samples

Quality Segment

agency

PCR, Immunomagnetic Separation (IMS), AOAC® (PTM), Health Canada, MicroVal

product line

Assurance GDS®

shelf life

6‑12 mo.

packaging

pkg of 100 tests

manufacturer/tradename

BioControl Systems 71009-100

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl GDS, for use with GDS Rotor-Gene

storage temp.

2-8°C

suitability

Listeria spp.

General description

Assurance® GDS, genetic detection system, for Listeria spp. Tq is an automated nucleic acid amplification system for the detection of Listeria in food and environmental samples. The proprietary PickPen® IMS-based sample preparation procedure quickly and effectively captures Listeria spp., separating the pathogen from inhibitory background materials ensuring valid PCR reactions for all sample types, including challenging food and environmental samples. Assurance® GDS PCR technology provides two additional levels of specificity through the use of both highly specific primers and probes to ensure accurate detection of Listeria spp.

Application

GDS, Genetic Detection System, for Listeria spp. Tq is an automated nucleic acid amplification system for the detection of Listeria species in foods, ingredients, and environmental samples.
PCR, Immunomagnetic Separation (IMS)

Features and Benefits

  • Results in 24 hours for environmental samples and 32 hours for product samples
  • Single-step enrichment reduces handling time
  • Features innovative PickPen® immunomagnetic separation (IMS) for additional specificity and faster time to results
  • Simplified sample prep does not require heating blocks or preparation of reagents
  • Proprietary Rotor-Gene® Q cycler is a rotory-based cycler that ensures uniform heating and cooling of all samples

Other Notes

Each 100 test kit includes:
  • Listeria spp. Amplification Tubes Tq (0.2 mL)
  • Listeria Concentration Reagent
  • Listeria Resuspension Buffer Tq
  • Wash Solution

Legal Information

AOAC is a registered trademark of AOAC International
Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Rotor-Gene is a registered trademark of Qiagen GmbH


Storage Class

10 - Combustible liquids

signalword

Danger

hcodes

Hazard Classifications

ED ENV 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library