Skip to Content
MilliporeSigma

73076BC

GDS 72 Well Rotor, RG Q Only

BioControl Systems, for use with GDS Rotor-Gene Thermocycler

Synonym(s):

Thermocycler Rotor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122409
NACRES:
NC.81
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene Thermocycler

storage temp.

room temp

General description

GDS 72 Well Rotor is an accessory to hold the amplification tubes (0.1 mL) containing samples in the GDS Rotor-Gene® Q thermocycler during amplification. GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The unique centrifugal rotary design of the Rotor-Gene® Q makes it the most precise and versatile thermocycler. Amplification tubes spin in a chamber of moving air, keeping all samples at precisely the same temperature during rapid thermal cycling thereby enabling uniform detection. When each tube aligns with the detection optics, the sample is illuminated, and the fluorescent signal is rapidly collected from a single, short optical pathway. This thermal and optical uniformity results in sensitive, precise, and fast PCR analysis.

Application

GDS 72 well Rotor is used with Assurance® GDS Rotor-Gene® Q thermocycler for the detection of pathogenic organisms in foods, ingredients, and environmental samples without the need for optimization of reaction and cycling conditions.

Features and Benefits

The rotor design delivers:
  • Tube-to-tube variation of ±0.02°C for uniform detection
  • Pre-loaded firmware specific to Assurance® GDS assays
  • Negligible equilibration times for fast run-cycling
  • Complete confidence in your results

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Rotor-Gene is a registered trademark of Qiagen GmbH




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library