Skip to Content
MilliporeSigma

73073BC

GDS USB Cable, RG Q Only

BioControl Systems, for use with GDS Rotor-Gene

Synonym(s):

USB cable

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122409
NACRES:
NC.81
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

General description

GDS USB Cable is an essential accessory of the Assurance® GDS Rotor-Gene Q Thermocycler. Assurance® GDS Rotor-Gene is used for the detection of pathogenic organisms in foods, ingredients, and environmental samples.

Application

GDS USB Cable is used to connect the Assurance® GDS Rotor-Gene Q to the communications port of the Assurance® GDS Q laptop.

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
BioControl is a registered trademark of BioControl Systems, Inc.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library