Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

580561

Millipore

TCEP, Neutral

A ready-to-use stable, neutral form of the popular disulfide-reducing agent TCEP, Hydrochloride.

Synonym(s):

TCEP, Neutral

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12ClNa4O6P
Molecular Weight:
374.57
UNSPSC Code:
12352128
NACRES:
NA.25

Quality Level

form

solid

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

General description

A ready-to-use stable, neutral form of the popular disulfide-reducing agent TCEP, Hydrochloride (Cat. No. 580560). Reconstitute each vial with 1 ml H2O to obtain a 500 mM solution.
A ready-to-use, stable, neutral form of TCEP hydrochloride (Cat. No. 580560), a disulfide reducing agent.

Application


  • Förster Resonance Energy Transfer Assay for Investigating the Reactivity of Thioesters in Biochemistry and Native Chemical Ligation.: This study utilizes TCEP in a FRET assay to explore thioester reactivity, providing insights into biochemical reactions and peptide synthesis applications (Gless et al., 2023).

  • Two-Dimensional Capillary Zone Electrophoresis-Mass Spectrometry: Intact mAb Charge Variant Separation Followed by Peptide Level Analysis Using In-Capillary Digestion.: TCEP is employed for in-capillary digestion to enhance protein separation and analysis, improving the characterization of monoclonal antibodies (Schlecht et al., 2023).

  • Silica Nanoparticle-Loaded Thermoresponsive Block Copolymer Vesicles: A New Post-Polymerization Encapsulation Strategy and Thermally Triggered Release.: TCEP is used in the synthesis of block copolymer vesicles, facilitating the encapsulation and controlled release of nanoparticles for biomedical applications (Czajka et al., 2022).

  • Thiol Catalysis of Selenosulfide Bond Cleavage by a Triarylphosphine.: This research demonstrates the use of TCEP in catalyzing selenosulfide bond cleavage, contributing to the understanding of thiol-based reduction mechanisms (Firstova et al., 2022).

  • A Novel PNGase Rc for Improved Protein N-Deglycosylation in Bioanalytics and Hydrogen-Deuterium Exchange Coupled With Mass Spectrometry Epitope Mapping Under Challenging Conditions.: TCEP is highlighted for its role in improving protein deglycosylation and mass spectrometry analysis, enhancing bioanalytical techniques (Gramlich et al., 2022).

Packaging

Packaged under inert gas

Warning

Toxicity: Corrosive (L)

Reconstitution

Following reconstitution, aliquot, flush with an inert gas, and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Reconstitute with 1 ml water to obtain a 500 mM solution.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Getz, E.B., et al. 1999. Anal. Biochem.273, 73.
Han, J.C. and Han, G.Y. 1994. Anal. Biochem.220, 5.
Burns, J.A., et al. 1991. J. Org. Chem.56, 2648.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service