Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22790

Sigma-Aldrich

2-Chloroacetamide

≥98.0% (HPLC)

Synonym(s):

Chloracetamide, alpha-Chloroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CONH2
CAS Number:
Molecular Weight:
93.51
Beilstein:
878191
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.05 mmHg ( 20 °C)

Assay

≥98.0% (HPLC)

form

solid

mp

116-118 °C (lit.)
118-120 °C

solubility

methanol: soluble 1 g/10 mL
absolute ethanol: 10 part(lit.)
water: soluble 10 parts(lit.)
diethyl ether: very slightly soluble(lit.)

SMILES string

NC(=O)CCl

InChI

1S/C2H4ClNO/c3-1-2(4)5/h1H2,(H2,4,5)

InChI key

VXIVSQZSERGHQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloroacetamide was used in the synthesis of the derivatives of 1,8-naphthyridine containing thiono groups. It was also used as a reagent for the synthesis of carboxamidomethyl esters as carboxyl protecting groups in peptide synthesis

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Martinez et al.
Tetrahedron Letters, 24, 5219-5219 (1983)
Jun Zhang et al.
Journal of agricultural and food chemistry, 59(9), 4614-4621 (2011-03-23)
A butachlor-degrading strain, designated FLY-8, was isolated from rice field soil and was identified as Paracoccus sp. Strain FLY-8 could degrade and utilize six chloroacetamide herbicides as carbon sources for growth, and the degradation rates followed the order alachlor >
Cleonice Rocha et al.
Environmental pollution (Barking, Essex : 1987), 152(1), 239-244 (2007-07-17)
Solid-phase microextraction coupled with gas chromatography-mass spectrometry (SPME-GC-MS) was used to analyze two triazine (atrazine and simazine) and three chloroacetamide herbicides (acetochlor, alachlor, and metolachlor) in water samples from a midwest US agricultural drainage ditch for two growing seasons. The
K Eszter Borbas et al.
Bioconjugate chemistry, 18(4), 1205-1212 (2007-06-23)
Targeted radiopharmaceuticals offer the possibility of improved tumor imaging and radiotherapy, with reduced side effects. A variety of monoclonal antibodies and antibody fragments have previously been successfully radiolabeled and used in diagnostic imaging and targeted radiotherapy of cancer. Many such
Yasmine Souissi et al.
Journal of chromatography. A, 1310, 98-112 (2013-09-10)
The photooxidation of acetochlor (a pesticide belonging to the acetamides group) using a polychromatic UV irradiation in ultrapure water was studied. This study reports the efficiency of mass spectrometry for the characterization of photodegradation products of acetochlor. Decompositions of protonated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service