Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

524390

Sigma-Aldrich

Phorbol 12,13-dibutyrate

≥99% (HPLC), solid (crystalline), PKC activator, Calbiochem

Synonym(s):

Phorbol-12,13-dibutyrate, PDBu, PKC Activator II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H40O8
CAS Number:
Molecular Weight:
504.61
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

product name

Phorbol-12,13-dibutyrate, Strong irritant for mouse skin, but only moderately active as a tumor promoter.

Quality Level

Assay

≥99% (HPLC)

form

solid (crystalline)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to pale yellow

shipped in

ambient

storage temp.

−20°C

InChI

1S/C28H40O8/c1-7-9-20(30)35-24-16(4)27(34)18(22-25(5,6)28(22,24)36-21(31)10-8-2)12-17(14-29)13-26(33)19(27)11-15(3)23(26)32/h11-12,16,18-19,22,24,29,33-34H,7-10,13-14H2,1-6H3/t16-,18+,19-,22-,24-,26-,27-,28-/m1/s1

InChI key

BQJRUJTZSGYBEZ-YVQNUNKESA-N

General description

Strong irritant for mouse skin, but only moderately active as a tumor promoter. Activates protein kinase C (PKC). Stimulates the phosphorylation of Na+-K+ ATPase, thereby inhibiting its activity. Promotes the expression of iNOS in cultured hepatocytes. PDBu can be diluted to 30 µM in aqueous solutions, so it is commonly used in binding studies or in applications requiring high concentrations. It is easier to remove PDBu than PMA from cells in culture.
Strong irritant for mouse skin, but only moderately active as a tumor promoter. Activates protein kinase C. Stimulates the phosphorylation of Na+,K+- ATPase, thereby inhibiting its activity. Promotes the expression of inducible NOS in cultured hepatocytes. Commonly used in binding studies or in applications requiring high concentrations of phorbol compounds.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Na+,K+- ATPase
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Preparation Note

Further dilute with aqueous buffers just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 6 months at -20°C. With frequent use, the DMSO stock solutions may accumulate water. The presence of water in the stock solution will accelerate the decomposition of the product, resulting in significant loss of activity in 2-3 months.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Fisone, G., et al. 1995. J. Biol. Chem. 270, 2427.
Lee, E.J., et al. 1994. Brain Res. 665, 127.
Critz, S.D., et al. 1993. J. Neurochem.60, 1175.
Iredale, P.A., et al. 1993. Biochem. Pharmacol.45, 611.
Critz, S.D., and Byrne J.H. 1992. J. Neurophys.68, 1079.
Hortelano, S., et al. 1992. J. Biol. Chem. 267, 24937.
Ward C.A., and Moffat, M.P. 1992. J. Mol. Cell. Cardiol.24, 937.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Fischer et al.
Journal of neuroinflammation, 18(1), 252-252 (2021-11-04)
Astrocytes participate in the local innate immune response of the central nervous system. In response to stress such as ischemia, activated cells release endogenous factors known as damage-associated molecular patterns (DAMPs). Self-extracellular RNA (eRNA) is such a ubiquitous alarm signal.
Nayara C Leite et al.
Cell reports, 32(2), 107894-107894 (2020-07-16)
Understanding the root causes of autoimmune diseases is hampered by the inability to access relevant human tissues and identify the time of disease onset. To examine the interaction of immune cells and their cellular targets in type 1 diabetes, we
An-Angela N Van et al.
The Journal of biological chemistry, 296, 100445-100445 (2021-02-23)
Within the AGC kinase superfamily, gene fusions resulting from chromosomal rearrangements have been most frequently described for protein kinase C (PKC), with gene fragments encoding either the C-terminal catalytic domain or the N-terminal regulatory moiety fused to other genes. Kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service