Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

368067-M

Millipore

Granzyme B Substrate VIII, Colorimetric

A colorimetric substrate for granzyme B (kcat/Km = 6.6 x 10⁴ M⁻¹s⁻¹).

Synonym(s):

Granzyme B Substrate VIII, Colorimetric, Ac-IEPD- pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H38N6O11
Molecular Weight:
634.63
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

storage temp.

−20°C

SMILES string

CCC(C)C(C(=O)NC(CCC(=O)O)C(=O)N1CCCC1C(=O)NC(CC(=O)O)C(=O)NC2=CC=C(C=C2)[N+](=O)[O-])NC(=O)C

InChI key

YXEIIOVAGGAMCZ-UHFFFAOYSA-N

General description

A colorimetric substrate for granzyme B (kcat/Km = 6.6 x 104 M-1s-1). Cleavage of pNA is monitored colorimetrically at ~405 nm (ε = 9,160 M-1cm-1).

Biochem/physiol Actions

Primary Target
Colorimetric substrate for granzyme B
kcat/Km = 6.6 x 104 M⁻¹ sec⁻¹ as substrate for granzyme B

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Ile-Glu-Pro-Asp-pNA

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Harris, J.L., et al. 1998. J. Biol. Chem. 273, 27364.
Thornberry, N.A., et al. 1997. J. Biol. Chem. 272, 17907.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service