Skip to Content
MilliporeSigma

368057

Granzyme B Substrate I, Colorimetric

Colorimetric substrate for the detection of granzyme B and caspase-8 activity.

Synonym(s):

Granzyme B Substrate I, Colorimetric, Ac-IETD- pNA, Caspase-8 Substrate I, Ac-IETD-pNA, Caspase-8 Substrate I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H38N6O12
Molecular Weight:
638.62
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
lyophilized solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Colorimetric substrate for detection of granzyme B. Cleavage is monitored colorimetrically at ~405 nm.
Colorimetric substrate for the detection of granzyme B and caspase-8 activity. Cleavage of pNA is monitored colorimetrically at ~405 nm.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Colorimetric substrate for the detection of granzyme B and caspase-8 activity
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ac-Ile-Glu-Thr-Asp-pNA
Koeplinger, K.A., et al. 2000. Protein Exp. Purif. 18, 378.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library