Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

368059-M

Millipore

Granzyme B Substrate II, Fluorogenic

A fluorogenic substrate for the detection of granzyme B and caspase-8 activity.

Synonym(s):

Granzyme B Substrate II, Fluorogenic, Z-IETD-AFC, Caspase-8 Substrate II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H42F3N5O13
Molecular Weight:
821.75
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 1 mg/mL

fluorescence

λex ~400 nm
λem ~505 nm

storage temp.

−20°C

General description

A fluorogenic substrate for the detection of granzyme B. Reaction can be monitored visually or quantitatively by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore (excitation maximum: ~400 nm; emission maximum: ~505 nm).

Biochem/physiol Actions

Primary Target
Fluorogenic substrate for the detection of granzyme B and caspase-8 activity

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Ile-Glu-Thr-Asp-AFC

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Karahashi, H., and Amano, F. 2000. Biol. Pharm. Bull. 23, 140.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service