Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6470

Sigma-Aldrich

N-Acetyl-Ile-Glu-Pro-Asp-p-nitroanilide

≥97% (HPLC), powder

Synonym(s):

Ac-IEPD-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H38N6O11
CAS Number:
Molecular Weight:
634.63
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

solubility

DMSO: soluble

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(C)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CC(O)=O)C(=O)Nc2ccc(cc2)[N+]([O-])=O

InChI

1S/C28H38N6O11/c1-4-15(2)24(29-16(3)35)27(42)31-19(11-12-22(36)37)28(43)33-13-5-6-21(33)26(41)32-20(14-23(38)39)25(40)30-17-7-9-18(10-8-17)34(44)45/h7-10,15,19-21,24H,4-6,11-14H2,1-3H3,(H,29,35)(H,30,40)(H,31,42)(H,32,41)(H,36,37)(H,38,39)/t15-,19-,20-,21-,24-/m0/s1

InChI key

YXEIIOVAGGAMCZ-NYMCBPKFSA-N

Related Categories

Amino Acid Sequence

NAc-Ile-Glu-Pro-Asp-pNA

Substrates

Chromogenic substrate for caspase 8 and granzyme B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Ho et al.
ACS synthetic biology, 6(8), 1484-1495 (2017-05-17)
Targeted therapies promise to increase the safety and efficacy of treatments against diseases ranging from cancer to viral infections. However, the vast majority of targeted therapeutics relies on the recognition of extracellular biomarkers, which are rarely restricted to diseased cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service