Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

249572

Sigma-Aldrich

Sodium acetylide suspension

95% (Purity of contained acetylide), 18 wt. % slurry in xylene: light mineral oil

Synonym(s):

Ethynyl sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CNa
CAS Number:
Molecular Weight:
48.02
Beilstein:
3587232
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95% (Purity of contained acetylide)

concentration

18 wt. % slurry in xylene: light mineral oil

SMILES string

[Na]C#C

InChI

1S/C2H.Na/c1-2;/h1H;

InChI key

SLBNQJAGYKQCKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium acetylide suspension has been used in the preparation of:
  • 2-ethynyladamantan-2-ol
  • apical-equatorial (ae) isomer of (Et4N)3[1-(2′-B10H9)-2-NH3B10H8]
  • trimethylsilylacetylene

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3 - Water-react 2

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service