Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46549

Millipore

Sabouraud Dextrose Agar

Chloramphenicol, lid, Lockable contact plate, plate diam. 55 mm, γ-irradiated, single packed of 20 plates, suitable for surface monitoring

Synonym(s):

SDA Contact plate, SDA with Chloramphenicol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

γ-irradiated

form

plate (ready-to-use)

feature

lid Lockable contact plate
ready-to-use

composition

Chloramphenicol

packaging

single packed of 20 plates

plate diam.

55 mm

pH

5.6 ( in H2O)

application(s)

cosmetics
food and beverages
pharmaceutical

surface monitoring

compatibility

stand alone

storage temp.

room temp

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Contact Agar + Chloramphenicol - RT+ is designed for the detection of the total count of yeasts and molds on dry surfaces and personnel in non-controlled environments. Sabouraud Dextrose Medium contains digests of animal tissues (peptones), which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose serves as the energy and carbon source. The low pH favors fungal growth and inhibits the growth of contaminating bacteria from samples. The addition of chloramphenicol further inhibits the growth of the accompanying bacterial flora. Additionally, the plate model (plus or “+”) is supplied with a lockable lid for safe transport after sampling and also aid in aerobic and anaerobic incubation of the plates.

Application

Sabouraud Dextrose Contact Agar + Chloramphenicol - RT+ is designed for the detection of the total count of yeasts and molds on dry surfaces and personnel in non-controlled environments and is suitable for Room Temperature storage.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service