Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.05438

Millipore

Sabouraud Dextrose Agar

GranuCult® prime, Dextrose 4 %, EP 5.6, USP 29, JP 4.05, granular, for yeasts, for molds, pack of 500 g and 5 kg

Synonym(s):

Fungus culture medium acc. to SABOURAUD, SABOURAUD 4% dextrose agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

EP 5.6
JP 4.05
USP 29

Quality Level

form

granular

composition

Dextrose, 4%

manufacturer/tradename

GranuCult® prime

packaging

pack of 500 g and 5 kg

technique(s)

microbiological culture: suitable

pH

5.6 (25 °C, 65 g/L in H2O, after autoclaving)

solubility

65 g/L

bulk density

680 kg/m3

application(s)

bioburden testing
industrial qc
microbiology
pharmaceutical

storage temp.

15-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud 4% Dextrose Agar is a complex, granulated, dehydrated culture medium for cultivation and isolation of yeasts and molds. It also serves as the absence test for Candida albicans. The high concentration of dextrose and the low pH promotes the growth, the formation of spores (Conidia and Sporangia), and the formation of pigments of yeasts and molds. Additionally, it inhibits the growth of bacteria.

Application

Sabouraud 4% Dextrose Agar is designed for the determination of the total count of yeasts and molds in non-sterile products.

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 5.4 - 5.8
Typical composition (g/litre): Peptone from casein5.0; Peptone from meat 5.0; D(+)Glucose 40.0; Agar-agar 15.0.
Growth (Trichophyton mentagrophytes ATCC 18748): fair to very good
Growth (Trichophyton rubrum ATCC 28188): fair to good
Growth (Trichophyton ajelloi ATCC 28454): fair to good
Growth (Microsporum gallinae ATCC 12108): fair to very good
Growth (Microsporum canis ATCC 36299): good to very good
Growth (Geotrichum candidum DSM 1240): good to very good
Growth (Penicillium commune ATCC 10428): good to very good
Incubation: 7 days; 28°C
Growth promotion test in accordance with the harmonised method of EP, USP and JP.
Inoculum on reference medium (Candida albicans ATCC 10231 (WDCM 00054)): 10 - 100
Inoculum on reference medium (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)): 10 - 100
Colony count (Candida albicans ATCC 10231 (WDCM 00054)):
Colony count (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)):
Recovery on test medium (Candida albicans ATCC 10231 (WDCM 00054)): ≥ 70 %
Recovery on test medium (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)): ≥ 50 %
Incubation: C. albicans and A. brasiliensis up to 5 daysat 20-25°C.
The information above is current at this time of publication and is subject to change without notice (except for customers holding a change control agreement with our company). The information/format can be altered at any time and inany way if internal company matters or Standard Regulationsdo say so.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danielle Aparecida Duarte Nunes et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 55(6), 650-660 (2020-02-19)
Bioremediation is one of the existing techniques applied for treating oil-contaminated soil, which can be improved by the incorporation of low-cost nutritional materials. This study aimed to assess the addition of two low-cost plant residues, sugarcane bagasse (SCB) and leaf

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service