Skip to Content
MilliporeSigma

1.46004

Tryptic Soy Agar - Ready-to-use Settle Plates

EP, USP, Settle Plate for long Incubation, aseptically filled, plate diam. 90 mm, suitable for air monitoring

Synonym(s):

CASO Agar, Casein Soybean Digest Agar, TSA, Tryptic Soy Agar Settle Plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106217
Plate diam.:
90 mm
Feature:
ready-to-use, Settle Plate for long Incubation
Sterility:
aseptically filled
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene plate

Quality Segment

agency

EP, USP

sterility

aseptically filled

form

plate (ready-to-use)

feature

ready-to-use, Settle Plate for long Incubation

packaging

pack of 120 plates, pack of 20 plates

plate diam.

90 mm

pH

7.3 ( in H2O)

application(s)

air monitoring, cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

15-25°C

suitability

bacteria, fungi

General description

Tryptic Soy Agar - in 90 mm settle plates with 30 mL filling volume for long incubation is designed for the detection of the total aerobic microbial count in the air via active or passive air monitoring as well as fingerprints of personnel in controlled environments.
TSA medium is used for the cultivation and isolation of a wide range of bacteria, yeasts, and molds. The formulation contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium.

Application

Tryptic Soy Agar in 90 mm settle plates is designed for the determination of the total aerobic microbial count in air via active or passive air monitoring as well as fingerprints of personnel in controlled environments.

Legal Information

MAS-100 is a registered trademark of MBV AG


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library