Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46201

Millipore

Sabouraud Dextrose Agar

Sodium thiosulphate, Lecithin, Tween®, Histidine, lid, ICR: non lockable contact plate, plate diam. 55 mm, sterile; irradiated, triple packed of 20 or 200 plates, suitable for surface monitoring

Synonym(s):

SDA contact plate, SDA, SDA with neutralizer, ICR contact plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate (ready-to-use)

feature

lid ICR: non lockable contact plate
ready-to-use

composition

Histidine
Lecithin
Sodium thiosulphate
Tween®

packaging

triple packed of 20 or 200 plates

plate diam.

55 mm

pH

5.6 ( in H2O)

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Contact Agar + LTHTh - ICR is designed for the determination of the total count of yeasts and molds on dry, sanitized surfaces and personnel in Clean rooms and Isolators. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH favours fungal growth. The media is supplemented with the neutralisers such as Lecithin, polysorbate (Tween®) 80, histidine, sodium thiosulfate and pyruvate. The plastic plate is of pink color to differentiate between TSA and SDA media.

Application

Sabouraud Dextrose Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The media is used for surface monitoring in isolators and clean rooms.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service