Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14543

Supelco

Phosphate Test, photometric

photometric, 0.05-5.00 mg/L (PO4-P), 0.2-15.3 mg/L (PO43-), 0.11-11.46 mg/L (P2O5), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Phosphate Cell Test, photometric, 0.05-5.00 mg/L (PO4-P), 0.2-15.3 mg/L (PO43-), 0.11-11.46 mg/L (P2O5), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

phosphate

measuring range

0.05-5.00 mg/L (PO4-P)
0.11-11.46 mg/L (P2O5)
0.2-15.3 mg/L (PO43-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Phosphomolybdenum blue)

storage temp.

15-25°C

General description

(o-phosphate and total phosphorous) Method: Photometric, PMB 0.05 - 5.00 mg/L (PO4-P), 0.2 - 15.3 mg/L (PO43-), 0.11 - 11.46 mg/L (P2O5), Spectroquant®
The method is analogous to EPA 365.2+3, APHA 4500-P E, and DIN EN ISO 6878.
The Spectroquant® Phosphate Cell Test allows the accurate quantification of phosphate. Elevated levels of phosphate might lead to eutrophication of water bodies, which is a risk to the environment. Therefore, the concentration of phosphate in drinking water and wastewater is regulated in many countries.
Further applications are the determination of phosphate in foods & beverages (e.g. beer) and in environmental and agricultural samples (e.g. soil).
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode), which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination of phosphate ions in manure samples using phosphorus molybdenum blue and Spectroquant® test kits.

Photometric determination of phosphate ions in manure samples using phosphorus molybdenum blue and Spectroquant® test kits.

Photometric determination of total phosphorus in plant material samples by phosphorus molybdenum blue method using Spectroquant® test kits.

Photometric determination of total phosphorus in plant material samples by phosphorus molybdenum blue method using Spectroquant® test kits.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service