Skip to Content
MilliporeSigma

Skip To

1.14541

COD Cell Test, photometric

photometric, 25-1500 mg/L (COD), Spectroquant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

COD Cell Test, photometric, 25-1500 mg/L (COD), Spectroquant®

product line

Spectroquant®

Quality Segment

usage

sufficient for 25 tests

analyte functional class(es)

chemical oxygen demand (COD)

measuring range

25-1500 mg/L (COD)

technique(s)

photometry: suitable

application(s)

aquaculture
disposal drainage water
electroplating
environmental
power plants
wastewater

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

detection method

photometric (Oxidation with chromosulfuric acid, determination as chromate)

storage temp.

15-25°C

General description

Method: Photometric, 25 - 1500 mg/L, Spectroquant®
The method corresponds to DIN ISO 15705 and is analogous to EPA 410.4, APHA 5220 D, and ASTM D1252-06 B.
The Chemical Oxygen Demand (COD) is a measure for the amount of oxygen consumed during the oxidation of organic matter, which gives a clear indication for the quality of a water source, it is for sure one of the most important parameters in (waste) water analysis. Our Spectroquant® COD Tests allow you to accurately quantify the of COD levels in your sample.
Select the right measurement range for your application needs from our wide range of Spectroquant® COD Tests.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant®Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode), which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.017971.146911.14555
detection method

photometric (Oxidation with chromosulfuric acid, determination as chromate)

detection method

photometric (Oxidation with chromosulfuric acid, determination as chromium(III))

detection method

photometric (Oxidation with chromosulfuric acid, determination as chromium(III))

detection method

photometric (Oxidation with chromosulfuric acid, determination as chromium(III))

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

measuring range

25-1500 mg/L (COD)

measuring range

5000-90000 mg/L (COD)

measuring range

300-3500 mg/L (COD)

measuring range

500-10000 mg/L (COD)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 600 (plus), for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 300 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

usage

sufficient for 25 tests

usage

sufficient for 25 tests

usage

sufficient for 25 tests

usage

25  tests

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C


Still not finding the right product?

Explore all of our products under COD Cell Test


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 1B - Repr. 1B - Skin Corr. 1A - STOT RE 2

target_organs

Kidney

Storage Class

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Will this assist with testing COD of my waste water?

    1 answer
    1. Yes, this product can assist with testing the COD of wastewater.

      Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/166/405/114541ug-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters