Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00798

Supelco

Phosphate Test

photometric, 1.0-100.0 mg/L (PO4-P), 3-307 mg/L (PO43-), 2-229 mg/L (P2O5), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests

specific analyte(s)

phosphate

measuring range

1.0-100.0 mg/L (PO4-P)
2-229 mg/L (P2O5)
3-307 mg/L (PO43-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Phosphomolybdenum blue)

storage temp.

15-25°C

General description

(o-phosphate) Method: photometric, PMB 1.0 - 100.0 mg/l PO₄-P 3 - 307 mg/l PO₄³⁻ 2 - 229 mg/l P₂O₅ Spectroquant®
The method is analogous to EPA 365.2+3, APHA 4500-P E, and DIN EN ISO 6878.
The Spectroquant® Phosphate Reagent Test allows the accurate quantification of phosphate. Elevated levels of phosphate might lead to eutrophication of water bodies, which is a risk to the environment. Therefore, the concentration of phosphate in drinking water and wastewater is regulated in many countries.
Further applications are the determination of phosphate in foods & beverages (e.g. beer) and in environmental and agricultural samples (e.g. soil).
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Application



  • Effects of Two Remineralizing Agents in Combination with Er:YAG and CO(2) Laser Irradiation on Microhardness of Demineralized Enamel: This study highlights the use of phosphate tests in dental research, specifically in measuring the efficacy of remineralizing agents combined with laser treatment to enhance enamel hardness, which is crucial for dental health and preventive care (Mapar M et al., 2024).


  • Zinc and Silicon Nano-Fertilizers Influence Ionomic and Metabolite Profiles in Maize to Overcome Salt Stress: This research utilizes phosphate tests in the study of plant physiology, specifically to gauge the efficacy of nano-fertilizers in enhancing phosphate uptake under salt stress conditions, providing insights into sustainable agricultural practices (Shoukat A et al., 2024).


  • Investigation of Mechanical Properties of Polymer-Infiltrated Tetrapodal Zinc Oxide in Different Variants: Applies phosphate testing in the field of materials science to assess the interaction of phosphate with novel composite materials, thereby informing the development of more durable and efficient materials for various industrial applications (Scherer F et al., 2024).


Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hoang Nhat Phong Vo et al.
Bioresource technology, 303, 122877-122877 (2020-02-07)
This study investigated the extracellular polymeric substance (EPS) and enzyme extrusion of Chlorella sp. using seven carbon sources and two salinities for potential pollutant co-metabolism. Results indicated that the levels of biomass, EPS and enzymes of microalgae cultured with glucose

Protocols

Photometric determination of phosphate ions in manure samples using phosphorus molybdenum blue and Spectroquant® test kits.

Photometric determination of phosphate ions in manure samples using phosphorus molybdenum blue and Spectroquant® test kits.

Photometric determination of phosphate ions in manure samples using phosphorus molybdenum blue and Spectroquant® test kits.

Photometric determination of phosphate ions in fertilizer samples using phosphorus molybdenum blue and Spectroquant® test kits.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service