Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14542

Supelco

Nitrate Test, photometric

photometric, 0.5-18.0 mg/L (NO3-N), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Nitrate Cell Test, photometric, 0.5-18.0 mg/L (NO3-N), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

nitrate

measuring range

0.5-18.0 mg/L (NO3-N)
2.2-79.7 mg/L (NO3-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Nitrospectral)

storage temp.

15-25°C

General description

Method: photometric 0.5 - 18.0 mg/l NO₃-N 2.2 - 79.7 mg/l NO₃⁻ Spectroquant®

This Spectroquant® Nitrate Cell Test allows the accurate quantification of nitrate in water samples. Nitrate is an essential nutrient required for the growth of animals and plants. However, elevated levels of nitrate can be a health risk, in particular for pregnant women and infants. Therefore, nitrate levels in water sources are regulated in many countries.
Further fields of applications are the analysis of foods & beverages (e.g. whey) and of environmental and agricultural samples (e.g. soil).
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A step-by-step procedure for the photometric determination of Nitrate in effluents using Spectroquant® test kits and photometer for a precise and quantitative analysis.

A step-by-step procedure for the photometric determination of Nitrate in effluents using Spectroquant® test kits and photometer for a precise and quantitative analysis.

Protocols

Learn about the photometric determination of nitrate ions in soil using Spectroquant® test kits and spectrophotometer.

Learn about the photometric determination of nitrate ions in compost using Spectroquant® test kits and spectrophotometer.

Learn about the photometric determination of nitrate ions in compost using Spectroquant® test kits and spectrophotometer.

Learn about the photometric determination of nitrate ions in compost using Spectroquant® test kits and spectrophotometer.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service