Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.11586

Supelco

Potassium hydroxide solution

c(KOH) = 0.5 mol/l (0.5 N), reag. USP, ready-to-use volumetric solution for titration, Titriplex®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21

Product Name

Potassium hydroxide solution, c(KOH) = 0.5 mol/l (0.5 N), Titripur®, reag. USP

Agency

reag. USP

Quality Level

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Acid-base reactions

concentration

0.5 M

technique(s)

titration: suitable

pH

>13.5 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[K+].[O-][H]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Voltammetric Determination: Potassium hydroxide solution in ethanol is used in voltammetric determination of antioxidants in various matrices. A recent study demonstrated its application in the voltammetric determination of the antioxidant 2,6-di-tert-butyl-p-cresol (T501) in transformer oil using a graphite electrode. This method enhances the sensitivity and accuracy of antioxidant detection, crucial for ensuring the stability and performance of oils and other industrial fluids (Zhou et al., Analytical Letters, 2021).

  • Oxidative Degradation Studies: The reagent is employed in oxidative degradation studies of organic compounds. For instance, it has been used in the oxidative degradation of piperazine in potassium hydroxide solution for CO2 absorption processes. This application is essential for developing efficient CO2 capture and storage technologies, contributing to environmental sustainability and climate change mitigation (Ochedi et al., SSRN Electronic Journal, 2022).

  • CO2 Absorption and Modeling: Potassium hydroxide solution in ethanol is used in experimental and modeling studies of CO2 absorption processes. Recent research has focused on the absorption of CO2 into potassium hydroxide aqueous solution, providing valuable data for optimizing industrial CO2 capture methods. This application is crucial for enhancing the efficiency and effectiveness of carbon capture technologies (Rastegar and Ghaemi, Heat and Mass Transfer, 2022).

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Analysis Note

Form liquid
Amount-of-substance concentration 0.4975 - 0.5025 mol/L
Measurement uncertainty ± 0.0015 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 84 potassium hydrogen phthalate) by means of volumetric standard potassium hydrogen phthalate (article number 1.02400), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service