Skip to Content
MilliporeSigma

1.05545

Potassium hydroxide solution

47%, for analysis EMSURE®

Synonym(s):

KOH Solution 47%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352305
NACRES:
NA.21
Concentration:
46.0-48.0% KOH
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Potassium hydroxide solution 47%, for analysis EMSURE®

Quality Segment

product line

EMSURE®

form

liquid

concentration

46.0-48.0% KOH

impurities

≤0.0001% As (Arsenic), ≤0.0003% Total nitrogen (N), ≤0.0005% Ca (Calcium), ≤0.0005% Cu (Copper), ≤0.0005% Fe (Iron), ≤0.0005% Heavy metals (as Pb), ≤0.0005% Mg (Magnesium), ≤0.0005% Ni (Nickel), ≤0.0005% Pb (Lead), ≤0.0005% Phosphate (PO4), ≤0.0005% Zn (Zinc), ≤0.001% Al (Aluminium), ≤0.002% Sulfate (SO4), ≤0.004% Chloride (Cl), ≤1.0% Carbonate (as K2CO3)

pH

>13.5 (20 °C in H2O)

storage temp.

2-30°C

Analysis Note

Assay (acidimetric): 46.0 - 48.0 %
Carbonate (as K₂CO₃): ≤ 1.0 %
Chloride (Cl): ≤ 0.004 %
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.002 %
Total nitrogen (N): ≤ 0.0003 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.001 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.0005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.0005 %
Ni (Nickel): ≤ 0.0005 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.0005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Potassium hydroxide solution 47%


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

wgk

WGK 1

Storage Class

8B - Non-combustible corrosive hazardous materials



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library